Novus Biologicals
Manufacturer Code:NB10093371
Catalog # NB10093371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-HMFSSKSPKPQE corresponding to internal region according to NP_000027.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: adenosine monophosphate deaminase 1 (isoform M); adenosine monophosphate deaminase 1 adenosine monophosphate deaminase 1 (isoform M) adenosine monophosphate deaminase-1 (muscle) AMP deaminase 1 AMP deaminase isoform M AMPD EC 3.5.4.6 MAD MADA Myoadenylate deaminase skeletal muscle AMPD; adenosine monophosphate deaminase-1 (muscle); AMP deaminase 1; AMP deaminase isoform M; AMPD; Myoadenylate deaminase; skeletal muscle AMPD
Gene Aliases: AMPD1; MAD; MADA; MMDD
UniProt ID: (Human) P23109
Entrez Gene ID: (Human) 270
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.