Invitrogen
Manufacturer Code:PA542267
Catalog # PIPA542267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human AMOTL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Angiomotin like 1 JEAP; Angiomotin-like protein 1; JEAP; junction-enriched and -associated protein; junction-enriched and associated protein
Gene Aliases: 2310010G08Rik; 2310067L22Rik; 4932416D09Rik; AMOTL1; JEAP; mFLJ00155
UniProt ID: (Human) Q8IY63, (Mouse) Q9D4H4
Entrez Gene ID: (Dog) 485122, (Human) 154810, (Rabbit) 100348712, (Mouse) 75723, (Guinea pig) 100720059
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.