Novus Biologicals
Manufacturer Code:NBP178210
Catalog # NBP178210
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 1034 and 1084 of human Angiomotin using the numbering given in entry NP_001106962.1 (GeneID 154796). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Angiomotin; angiomotin angiomotin p80 isoform KIAA1071angiomotin p130 isoform; angiomotin p130 isoform; angiomotin p80 isoform
Gene Aliases: AMOT; KIAA1071
UniProt ID: (Human) Q4VCS5
Entrez Gene ID: (Human) 154796
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.