Proteintech
Manufacturer Code:212781AP150UL
Catalog # 50-173-3347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AMIGO3 Fusion Protein Ag15790 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adhesion molecule with Ig-like domain 3; ALI3 Alivin 3 AMIGO 3 AMIGO3 Amphoterin induced protein 3 KIAA1851; alivin 3; Alivin-3; AMIGO-3; AMIGO3; amigo3 {ECO:0000312|EMBL:AAO48952.1, ECO:0000312|RGD:631413}; amphoterin induced gene and ORF 3; amphoterin-induced gene and ORF 3; Amphoterin-induced protein 3; transmembrane protein AMIGO3
Gene Aliases: ALI3; AMIGO-3; AMIGO3; E430002N15Rik; KIAA1851; mKIAA1851; UNQ6084/PRO20089
UniProt ID: (Human) Q86WK7, (Rat) Q80ZD5, (Mouse) Q8C2S7
Entrez Gene ID: (Human) 386724, (Rat) 316003, (Mouse) 320844
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.