Invitrogen
Manufacturer Code:PA5103183
Catalog # PIPA5103183
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AIFM2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5430437E11Rik AIFM2 AMID apoptosis inducing factor mitochondria associated 2 apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death apoptosis-inducing factor 2 Apoptosis-inducing factor homologous mitochondrion-associated inducer of death apoptosis-inducing factor mitochondrion-associated 2 apoptosis-inducing factor mitochondrion-associated 2 apoptosis-inducing factor-like mitchondrion-associated inducer of death apoptosis-inducing factor-like mitochondrion-associated inducer of death D730001I10Rik MBP2 MBPH MGC126662 MGC141971 p53-responsive gene 3 protein PRG3 RP11-367H5.2; AMID; apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death; apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death; apoptosis-inducing factor 2; Apoptosis-inducing factor homologous mitochondrion-associated inducer of death; apoptosis-inducing factor, mitochondrion-associated, 2; apoptosis-inducing factor-like mitchondrion-associated inducer of death; apoptosis-inducing factor-like mitochondrion-associated inducer of death; Ferroptosis suppressor protein 1; FSP1; p53-responsive gene 3 protein
Gene Aliases: 5430437E11Rik; AIFM2; AMID; D730001I10Rik; PRG3
UniProt ID: (Human) Q9BRQ8, (Mouse) Q8BUE4
Entrez Gene ID: (Human) 84883, (Mouse) 71361
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.