Novus Biologicals
Manufacturer Code:NBP258117
Catalog # NBP258117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MLVDMKDSFHHNVAALRASVETGFAKKTFISYSVTFKDNFRQGLVVGIDLKNQMVLLQGGEALPFSHLILATGSTGPFPGKFNEVSSQQAAIQAYEDMVRQVQRSRFIVVVGGGSAGVEMAAEIKTEYPEKEVTLIHSQVALAD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMID; AMID apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death apoptosis-inducing factor 2 Apoptosis-inducing factor homologous mitochondrion-associated inducer of death apoptosis-inducing factor mitochondrion-associated 2 Apoptosis-inducing factor-like mitochondrion-associated inducer of death DKFZp686L1298 FLJ14497 p53-responsive gene 3 protein PRG3apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer ofdeath RP11-367H5.2; apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death; apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death; Apoptosis-inducing factor homologous mitochondrion-associated inducer of death; apoptosis-inducing factor, mitochondrion-associated, 2; Ferroptosis suppressor protein 1; FSP1; p53-responsive gene 3 protein
Gene Aliases: AIFM2; AMID; PRG3
UniProt ID: (Human) Q9BRQ8
Entrez Gene ID: (Human) 84883
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.