Novus Biologicals
Manufacturer Code:NB200170
Catalog # NB200170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to residues 170-185 [VTLIHSQVALADKELL] of the 41 kDa human PRG3 protein. This sequence is 87% identical in mouse. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMID; AMID apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death apoptosis-inducing factor 2 Apoptosis-inducing factor homologous mitochondrion-associated inducer of death apoptosis-inducing factor mitochondrion-associated 2 Apoptosis-inducing factor-like mitochondrion-associated inducer of death DKFZp686L1298 FLJ14497 p53-responsive gene 3 protein PRG3apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer ofdeath RP11-367H5.2; apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death; apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death; Apoptosis-inducing factor homologous mitochondrion-associated inducer of death; apoptosis-inducing factor, mitochondrion-associated, 2; Ferroptosis suppressor protein 1; FSP1; p53-responsive gene 3 protein
Gene Aliases: AIFM2; AMID; PRG3
UniProt ID: (Human) Q9BRQ8
Entrez Gene ID: (Human) 84883
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.