Novus Biologicals
Manufacturer Code:NBP214286
Catalog # NBP214286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: KKTCGNKSSVNSTVLVKNTKKTNPEIKEKPCHFERCEGEKHIYSPIIVRE VIEEEEPSEKSEATYMTMHPVWPSLRSDRNNSL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adhesion molecule AMICA; Adhesion molecule interacting with CXADR antigen 1; adhesion molecule interacts with CXADR antigen 1 AMICA Gm638; adhesion molecule, interacts with CXADR antigen 1; Dendritic cell-specific protein CREA7-1; dendritic-cell specific protein CREA7-1; dendritic-cell specific protein CREA7-4; Junctional adhesion molecule-like
Gene Aliases: AMICA; AMICA1; CREA7-1; CREA7-4; Gm638; JAML; UNQ722/PRO1387
UniProt ID: (Human) Q86YT9
Entrez Gene ID: (Human) 120425
Molecular Function: cell adhesion molecule ion channel sodium channel transporter voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.