Novus Biologicals
Manufacturer Code:NBP257231
Catalog # NBP257231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DSLENSPPGDDCLYDLHGRSSEMFDPFLNFEPFLSSRPPGAMETEEERLVTIQKQLLYWELRREQLEAQEARAREAHARE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenomatous polyposis coli membrane recruitment 1; Adenomatous Polyposis Coli Membrane Recruitment 1 Amer1 APC Membrane Recruitment Protein 1 FAM123B Family With Sequence Similarity 123B OSCS Protein FAM123B RP11-403E24.2 Wilms Tumor Gene On The X Chromosome Protein Wilms Tumor On The X WTX; Amer1; APC membrane recruitment protein 1; family with sequence similarity 123B; Protein FAM123B; RP11-403E24.2; Wilms tumor gene on the X chromosome protein; Wilms tumor on the X
Gene Aliases: AMER1; FAM123B; OSCS; WTX
UniProt ID: (Human) Q5JTC6
Entrez Gene ID: (Human) 139285
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.