Novus Biologicals
Manufacturer Code:NBP215373
Catalog # NBP215373
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human AMELX. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIH1 ALGN amelogenin (amelogenesis imperfecta 1 X-linked) amelogenin (X chromosome amelogenesis imperfecta 1) amelogenin X isoform amelogenin X-linked AMGAMGL AMGXamelogenesis imperfecta 1; amelogenesis imperfecta 1; amelogenin (amelogenesis imperfecta 1, X-linked); amelogenin (X chromosome, amelogenesis imperfecta 1); Amelogenin, X isoform
Gene Aliases: AI1E; AIH1; ALGN; AMELX; AMG; AMGL; AMGX
UniProt ID: (Human) Q99217
Entrez Gene ID: (Human) 265
Molecular Function: extracellular matrix protein extracellular matrix structural protein structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.