Novus Biologicals
Manufacturer Code:NBP299173R
Catalog # NB058670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human AMDHD2. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amidohydrolase domain containing 2 Amidohydrolase domain-containing protein 2 CGI-14 EC 3.5.1.25 GlcNAc 6-P deacetylase putative N-acetylglucosamine-6-phosphate deacetylase; Amidohydrolase domain-containing protein 2; GlcNAc 6-P deacetylase; N-acetylglucosamine-6-phosphate deacetylase; putative N-acetylglucosamine-6-phosphate deacetylase
Gene Aliases: AMDHD2; CGI-14
UniProt ID: (Human) Q9Y303
Entrez Gene ID: (Human) 51005
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.