Invitrogen
Manufacturer Code:PA569894
Catalog # PIPA569894
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human AMDHD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amidohydrolase domain-containing protein 1; amidohydrolase domain-containing protein 1 probable imidazolonepropionase; Probable imidazolonepropionase
Gene Aliases: 1300019J08Rik; AMDHD1; HMFT1272; RGD1308879
UniProt ID: (Human) Q96NU7, (Bovine) A5PJV3, (Mouse) Q9DBA8
Entrez Gene ID: (Horse) 100065538, (Human) 144193, (Rabbit) 100357212, (Bovine) 505315, (Rat) 299735, (Guinea pig) 100721087, (Mouse) 71761
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.