Novus Biologicals
Manufacturer Code:NBP154975
Catalog # NBP154975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to AMD1(adenosylmethionine decarboxylase 1) The peptide sequence was selected from the N terminal of AMD1. Peptide sequence MGRMNSDCWYLYTLDFPESRVISQPDQTLEILMSELDPAVMDQFYMKDGV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosylmethionine decarboxylase 1 AdoMetDC AMD DKFZp313L1234 EC 4.1.1.50 S-adenosylmethionine decarboxylase 1 S-adenosylmethionine decarboxylase proenzyme SAMDCFLJ26964; AdoMetDC; S-adenosylmethionine decarboxylase 1; S-adenosylmethionine decarboxylase alpha chain; S-adenosylmethionine decarboxylase beta chain; S-adenosylmethionine decarboxylase proenzyme
Gene Aliases: ADOMETDC; AMD; AMD1; SAMDC
UniProt ID: (Human) P17707
Entrez Gene ID: (Human) 262
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.