Invitrogen
Manufacturer Code:MA514793
Catalog # PIMA514793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | R.16.1 |
Immunogen | Purified truncated recombit human AMACR-His fusion protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2-arylpropionyl-CoA epimerase; 2-methylacyl-CoA racemase; 2-methylacyl-CoA racemase OTTHUMP00000219978 OTTHUMP00000219979; Alpha-methylacyl-CoA racemase; alpha-methylacyl-Coenzyme A racemase
Gene Aliases: AMACR; AMACRD; CBAS4; Macr1; RACE; RM
UniProt ID: (Human) Q9UHK6, (Mouse) O09174
Entrez Gene ID: (Human) 23600, (Mouse) 17117
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.