OriGene
Manufacturer Code:TA800852S
Catalog # 50-167-8592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F10 |
Immunogen | Full length human recombit protein of human AMACR produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2-methylacyl-CoA racemase; Alpha-methylacyl-CoA racemase; AMACRD CBAS4 P504S RACE RM
Gene Aliases: AMACR; AMACRD; CBAS4; RACE; RM
UniProt ID: (Human) Q9UHK6
Entrez Gene ID: (Human) 23600
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.