Invitrogen
Manufacturer Code:PA5102360
Catalog # PIPA5102360
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AMACR corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-arylpropionyl-CoA epimerase; 2-arylpropionyl-CoA epimerase 2-methylacyl-CoA racemase Alpha-methylacyl-CoA racemase alpha-methylacyl-Coenzyme A racemase Amacr AMACRD amcr CBAS4 Da1-8 Macr1 Marc1 OTTHUMP00000219978 OTTHUMP00000219979 RACE RM; 2-methylacyl-CoA racemase; Alpha-methylacyl-CoA racemase; alpha-methylacyl-Coenzyme A racemase
Gene Aliases: AMACR; AMACRD; CBAS4; Da1-8; Macr1; Marc1; RACE; RM
UniProt ID: (Human) Q9UHK6, (Mouse) O09174
Entrez Gene ID: (Human) 23600, (Rat) 25284, (Mouse) 17117
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.