Invitrogen
Manufacturer Code:PA131261
Catalog # PIPA131261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunomicroscopy (IM) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide GAAVLRRLCKRSDVLLEPFR corresponding to amino acids 62-81 of Human AMCR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-arylpropionyl-CoA epimerase; 2-methylacyl-CoA racemase; Alpha-methylacyl-CoA racemase; alpha-methylacyl-Coenzyme A racemase; CBAS4 RACE RM
Gene Aliases: AMACR; AMACRD; CBAS4; Da1-8; Macr1; Marc1; RACE; RM
UniProt ID: (Human) Q9UHK6, (Mouse) O09174
Entrez Gene ID: (Human) 23600, (Rat) 25284, (Mouse) 17117
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.