Invitrogen
Manufacturer Code:MA135004
Catalog # 01-677-046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | V17.1 |
Immunogen | Alkaline phophatase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alkaline phosphatase 2; alkaline phosphatase 2, liver; Alkaline phosphatase liver/bone/kidney isozyme; alkaline phosphatase precursor (EC 3.1.3.2); alkaline phosphatase tissue non-specific isoform; alkaline phosphatase tissue non-specific isoform/TNS-AP; alkaline phosphatase, liver/bone/kidney; Alkaline phosphatase, tissue-nonspecific isozyme; AP-TNAP; AP-TNAP FLJ40094 FLJ93059 HOPS MGC161443 MGC167935 TNAP TNSALP; liver/bone/kidney isozyme; liver/bone/kidney-type alkaline phosphatase; tissue-nonspecific alkaline phosphatase; tissue-nonspecific ALP; Tissue-nonspecific ALP alkaline phosphatase; TNSALP
Gene Aliases: Akp-2; Akp2; ALPL; AP-TNAP; APTNAP; HOPS; PHOA; TNAP; TNS-AP; TNSALP
UniProt ID: (Bovine) P09487, (Chicken) Q92058, (Human) P05186, (Cat) Q29486, (Rat) P08289, (Mouse) P09242
Entrez Gene ID: (Bovine) 280994, (Chicken) 396317, (Human) 249, (Cat) 727689, (Rat) 25586, (Mouse) 11647
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.