Abnova
Manufacturer Code:MAB19541
Catalog # 89-336-066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | DBF-1 |
Immunogen | A synthetic peptide corresponding to human ALPL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C for one year. After reconstitution at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase, tissue-nonspecific isozyme; AP-TNAP; AP-TNAP FLJ40094 FLJ93059 HOPS MGC161443 MGC167935 TNAP TNSALP; liver/bone/kidney-type alkaline phosphatase; tissue-nonspecific ALP
Gene Aliases: ALPL; AP-TNAP; APTNAP; HOPS; TNAP; TNSALP
UniProt ID: (Human) P05186
Entrez Gene ID: (Human) 249
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.