Abnova
Manufacturer Code:MAB14793
Catalog # 89-034-876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ALPL/597 |
Immunogen | Recombinant protein corresponding to full length human ALPL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 to -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase, tissue-nonspecific isozyme; AP-TNAP; FLJ40094; FLJ93059; HOPS; liver/bone/kidney-type alkaline phosphatase; MGC161443; MGC167935; tissue-nonspecific ALP; TNAP; TNSALP
Gene Aliases: ALPL; AP-TNAP; APTNAP; HOPS; TNAP; TNSALP
UniProt ID: (Human) P05186
Entrez Gene ID: (Human) 249
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.