Invitrogen
Manufacturer Code:MA524845
Catalog # PIMA524845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 3G2 |
Immunogen | Recombit ALPL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Alkaline phosphatase 2; alkaline phosphatase 2, liver; Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase tissue-nonspecific isozyme AP-TNAP TNSALP Alkaline phosphatase liver/bone/kidney isozyme ALPL; Alkaline phosphatase, tissue-nonspecific isozyme; AP-TNAP; liver/bone/kidney-type alkaline phosphatase; tissue-nonspecific alkaline phosphatase; tissue-nonspecific ALP; Tissue-nonspecific ALP alkaline phosphatase; TNSALP
Gene Aliases: Akp-2; Akp2; ALPL; AP-TNAP; APTNAP; HOPS; PHOA; TNAP; TNSALP
UniProt ID: (Human) P05186, (Rat) P08289, (Mouse) P09242
Entrez Gene ID: (Human) 249, (Rat) 25586, (Mouse) 11647
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.