Abnova
Manufacturer Code:PAB3138
Catalog # 89-130-851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human ALPL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ALPL.
For Research Use Only
Protein Aliases: Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase, tissue-nonspecific isozyme; AP-TNAP; FLJ40094; FLJ93059; HOPS; liver/bone/kidney-type alkaline phosphatase; MGC161443; MGC167935; tissue-nonspecific ALP; TNAP; TNSALP
Gene Aliases: ALPL; AP-TNAP; APTNAP; HOPS; TNAP; TNSALP
UniProt ID: (Human) P05186
Entrez Gene ID: (Human) 249
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.