Invitrogen
Manufacturer Code:PA5103350
Catalog # PIPA5103350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ALPK3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha kinase 3 alpha-kinase 3 alpha-protein kinase 3 ALPK3 AW319487 D330016D04 KIAA1330 MAK MIDORI mKIAA1330 muscle alpha-kinase muscle alpha-protein kinase myocyte induction differentiation originator myocytic induction/differentiation originator; alpha-kinase 3; Alpha-protein kinase 3; alpk3; muscle alpha-kinase; Muscle alpha-protein kinase; myocyte induction differentiation originator; myocytic induction/differentiation originator
Gene Aliases: ALPK3; AW319487; D330016D04; KIAA1330; MAK; MIDORI; mKIAA1330
UniProt ID: (Human) Q96L96
Entrez Gene ID: (Human) 57538, (Mouse) 116904
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.