Invitrogen
Manufacturer Code:PA5104538
Catalog # PIPA5104538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ALPK2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha kinase 2 alpha-kinase 2 alpha-protein kinase 2 Alpk2 Gm549 HAK heart alpha-kinase heart alpha-protein kinase RGD1564431 RNF18; alpha-kinase 2; Alpha-protein kinase 2; alpk2; heart alpha-kinase; Heart alpha-protein kinase
Gene Aliases: ALPK2; Gm549; HAK; RGD1564431
UniProt ID: (Human) Q86TB3, (Mouse) Q91ZB0
Entrez Gene ID: (Human) 115701, (Mouse) 225638, (Rat) 498875
Molecular Function:
Hsp70 family chaperone
chaperone
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.