Proteintech
Manufacturer Code:191071AP150UL
Catalog # 50-173-2343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALPK1 Fusion Protein Ag5320 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 8430410J10Rik alpha kinase 1 Alpha protein kinase 1 ALPK1 Chromosome 4 kinase KIAA1527 LAK; alpha-kinase 1; Alpha-protein kinase 1; Chromosome 4 kinase; lymphocyte alpha-kinase; Lymphocyte alpha-protein kinase
Gene Aliases: 8430410J10Rik; ALPK1; KIAA1527; LAK; RGD1304613
UniProt ID: (Human) Q96QP1
Entrez Gene ID: (Human) 80216, (Mouse) 71481, (Rat) 310879
Molecular Function:
Hsp70 family chaperone
chaperone
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.