Invitrogen
Manufacturer Code:PA569994
Catalog # PIPA569994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse ALP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: asparaginase like 1; asparaginase-like 1; asparaginase-like 1 protein; Asparaginase-like protein 1; Asparaginase-like sperm autoantigen; ASRGL1 ALP asparaginase like 1 ALP1 FLJ22316 asparaginase-like 1 protein; Beta-aspartyl-peptidase; Glial asparaginase; Isoaspartyl dipeptidase; Isoaspartyl peptidase/L-asparaginase; Isoaspartyl peptidase/L-asparaginase alpha chain; Isoaspartyl peptidase/L-asparaginase beta chain; L-asparaginase; L-asparagine amidohydrolase; testis secretory sperm-binding protein Li 242mP
Gene Aliases: 2410004D18Rik; ALP; ALP1; ASRGL1; AU040643; AW060375; CRASH; Gliap; Hiob; Hiob2; zgc:103568
UniProt ID: (Human) Q7L266, (Rat) Q8VI04, (Mouse) Q8C0M9, (Zebrafish) Q5BKW9
Entrez Gene ID: (Human) 80150, (Rabbit) 100340867, (Horse) 100146576, (Rat) 246307, (Mouse) 66514, (Guinea pig) 100713636, (Zebrafish) 541402
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.