Invitrogen
Manufacturer Code:PA5106391
Catalog # PIPA5106391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ASRGL1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410004D18Rik ALP ALP1 asparaginase like 1 asparaginase-like 1 asparaginase-like 1 protein asparaginase-like protein 1 Asparaginase-like sperm autoantigen ASRGL1 AU040643 AW060375 beta-aspartyl-peptidase CRASH FLJ22316 Glial asparaginase Gliap Hiob Hiob2 Isoaspartyl dipeptidase Isoaspartyl peptidase/L-asparaginase Isoaspartyl peptidase/L-asparaginase alpha chain Isoaspartyl peptidase/L-asparaginase beta chain L-asparaginase L-asparagine amidohydrolase testis secretory sperm-binding protein Li 242mP; asparaginase-like 1; asparaginase-like 1 protein; Asparaginase-like protein 1; Asparaginase-like sperm autoantigen; Beta-aspartyl-peptidase; Glial asparaginase; Isoaspartyl dipeptidase; Isoaspartyl peptidase/L-asparaginase; Isoaspartyl peptidase/L-asparaginase alpha chain; Isoaspartyl peptidase/L-asparaginase beta chain; L-asparaginase; L-asparagine amidohydrolase; testis secretory sperm-binding protein Li 242mP
Gene Aliases: 2410004D18Rik; ALP; ALP1; ASRGL1; AU040643; AW060375; CRASH; Gliap; Hiob; Hiob2
UniProt ID: (Human) Q7L266, (Rat) Q8VI04, (Mouse) Q8C0M9
Entrez Gene ID: (Human) 80150, (Rat) 246307, (Mouse) 66514
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.