Invitrogen
Manufacturer Code:PA578762
Catalog # PIPA578762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human ALOX5 recombit protein (Position: A120-R483). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5 - Lipoxygenase; 5-lipoxygenase; 5-LO; Arachidonate 5-lipoxygenase; Arachidonate 5-lipoxygenase 5-LO 5-lipoxygenase ALOX5 LOG5; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; leukotriene A4 synthase; LOX-5; Polyunsaturated fatty acid 5-lipoxygenase
Gene Aliases: 5-LO; 5-LOX; 5LO; 5LPG; 5LX; AI850497; ALOX5; F730011J02; LOG5; LOX5A
UniProt ID: (Human) P09917, (Rat) P12527, (Mouse) P48999
Entrez Gene ID: (Human) 240, (Rat) 25290, (Mouse) 11689
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.