Invitrogen
Manufacturer Code:PA564932
Catalog # PIPA564932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human ALOX15B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15-lipoxygenase 2; 15-LOX-2; 15-LOX-2 15-LOX-B 15S-lipoxygenase arachidonate 15-lipoxygenase 2 arachidonate 15-lipoxygenase B arachidonate 15-lipoxygenase type II arachidonate 15-lipoxygenase second type arachidonate omega(6) lipoxygenase; 15-LOX-B; 15S-lipoxygenase; arachidonate 15-lipoxygenase 2; Arachidonate 15-lipoxygenase B; Arachidonate 15-lipoxygenase type II; arachidonate 15-lipoxygenase, second type; arachidonate omega(6) lipoxygenase; Linoleate 13-lipoxygenase 15-LOb; Polyunsaturated fatty acid lipoxygenase ALOX15B
Gene Aliases: 15-LOX-2; ALOX15B
UniProt ID: (Human) O15296
Entrez Gene ID: (Human) 247
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.