OriGene
Manufacturer Code:CF504250
Catalog # 50-166-2746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G8 |
Immunogen | Full length human recombit protein of human ALOX15 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 12-LOX; 12-LOX 15-LOX 15-LOX-1 LOG15; 12/15-lipoxygenase; 12/15-LOX; 15-lipooxygenase-1; 15-LOX; Arachidonate 12-lipoxygenase, leukocyte-type; Arachidonate 15-lipoxygenase; Arachidonate omega-6 lipoxygenase; Hepoxilin A3 synthase Alox15; Linoleate 13S-lipoxygenase; Polyunsaturated fatty acid lipoxygenase ALOX15
Gene Aliases: 12-LOX; 15-LOX; 15-LOX-1; 15LOX-1; Alox12; Alox12l; ALOX15; LOG15
UniProt ID: (Human) P16050, (Rat) Q02759
Entrez Gene ID: (Human) 246, (Dog) 489458, (Rat) 81639
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.