Invitrogen
Manufacturer Code:PA523608
Catalog # PIPA523608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 622-652 amino acids from the C-terminal region of human ALOX12B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 12R-lipoxygenase; 12R-LOX; 12R-LOX 12R-lipoxygenase arachidonate 12-lipoxygenase 12R-type epidermis-type lipoxygenase 12; Arachidonate 12-lipoxygenase, 12R-type; Epidermis-type lipoxygenase 12
Gene Aliases: 12R-LOX; ALOX12B; ARCI2
UniProt ID: (Human) O75342
Entrez Gene ID: (Human) 242
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.