Invitrogen
Manufacturer Code:PA578760
Catalog # PIPA578760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human 12 Lipoxygenase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 12(S)-lipoxygenase; 12S-lipoxygenase; 12S-LOX; Arachidonate (12S)-lipoxygenase; Arachidonate (15S)-lipoxygenase; arachidonate 12-lipoxygenase; Arachidonate 12-lipoxygenase 12S-type 12S-LOX 12S-lipoxygenase Lipoxin synthase 12-LO Platelet-type lipoxygenase 12 ALOX12 12LO LOG12; arachidonate 12-lipoxygenase, 12S-type; Linoleate (13S)-lipoxygenase; Lipoxin synthase 12-LO; P-12LO; platelet 12-LOX; platelet-type 12-lipoxygenase; Platelet-type lipoxygenase 12; Polyunsaturated fatty acid lipoxygenase ALOX12
Gene Aliases: 12-LO; 12-LOX; 12LO; 12S-LOX; 9930022G08Rik; ALOX12; Alox12p; LOG12; P-12LO
UniProt ID: (Human) P18054, (Mouse) P39655
Entrez Gene ID: (Human) 239, (Rat) 287454, (Mouse) 11684
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.