Invitrogen
Manufacturer Code:PA5103501
Catalog # PIPA5103501
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ALKBH7 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310045B01Rik 2510008E23Rik Abh7 alkB homolog 7 alkB alkylation repair homolog 7 alkB alkylation repair homolog 7 (E. coli) Alkbh7 Alkylated DNA repair protein alkB homolog 7 alpha-ketoglutarate-dependent dioxygenase alkB homolog 7 mitochondrial probable alpha-ketoglutarate-dependent dioxygenase ABH7 SPATA11 spermatogenesis associated 11 spermatogenesis cell proliferation related protein spermatogenesis cell proliferation-related protein Spermatogenesis-associated protein 11 UNQ6002 UNQ6002/PRO34564; alkB, alkylation repair homolog 7; Alkylated DNA repair protein alkB homolog 7; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 7, mitochondrial; probable alpha-ketoglutarate-dependent dioxygenase ABH7; spermatogenesis associated 11; spermatogenesis cell proliferation related protein; Spermatogenesis cell proliferation-related protein; Spermatogenesis-associated protein 11
Gene Aliases: 2310045B01Rik; 2510008E23Rik; ABH7; ALKBH7; SPATA11; UNQ6002; UNQ6002/PRO34564
UniProt ID: (Human) Q9BT30, (Mouse) Q9D6Z0
Entrez Gene ID: (Human) 84266, (Mouse) 66400
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.