Invitrogen
Manufacturer Code:PA5100873
Catalog # PIPA5100873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ALKBH5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABH5 AlkB family member 5 RNA demethylase alkB homolog 5 RNA demethylase alkB alkylation repair homolog 5 alkB alkylation repair homolog 5 (E. coli) Alkbh5 Alkylated DNA repair protein alkB homolog 5 alpha-ketoglutarate-dependent dioxygenase alkB homolog 5 AW050020 AW215868 E130207K11 OFOXD OFOXD1 oxoglutarate and iron-dependent oxygenase domain containing probable alpha-ketoglutarate-dependent dioxygenase ABH5 RGD1309496 RNA demethylase ALKBH5; AlkB family member 5, RNA demethylase; alkB, alkylation repair homolog 5; Alkylated DNA repair protein alkB homolog 5; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 5; oxoglutarate and iron-dependent oxygenase domain containing; probable alpha-ketoglutarate-dependent dioxygenase ABH5; RNA demethylase ALKBH5
Gene Aliases: ABH5; ALKBH5; AW050020; AW215868; E130207K11; OFOXD; OFOXD1; RGD1309496
UniProt ID: (Human) Q6P6C2, (Mouse) Q3TSG4, (Rat) D3ZKD3
Entrez Gene ID: (Human) 54890, (Mouse) 268420, (Rat) 303193
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.