Novus Biologicals
Manufacturer Code:NBP292048100UL
Catalog # NB025988
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 245-394 of human ALKBH5 (NP_060228.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABH5OFOXD alkB alkylation repair homolog 5 (E. coli) Alkylated DNA repair protein alkB homolog 5 EC 1.14.11.- FLJ20308 OFOXD1 oxoglutarate and iron-dependent oxygenase domain containing probable alpha-ketoglutarate-dependent dioxygenase ABH5; AlkB family member 5, RNA demethylase; alkB, alkylation repair homolog 5; Alkylated DNA repair protein alkB homolog 5; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 5; oxoglutarate and iron-dependent oxygenase domain containing; probable alpha-ketoglutarate-dependent dioxygenase ABH5; RNA demethylase ALKBH5
Gene Aliases: ABH5; ALKBH5; OFOXD; OFOXD1
UniProt ID: (Human) Q6P6C2
Entrez Gene ID: (Human) 54890
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.