Invitrogen
Manufacturer Code:PA5113466
Catalog # PIPA5113466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Alpha-ketoglutarate-dependent dioxygenase alkB homolog 4 protein (1-115AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010004B12Rik ABH4 Abj4 alkB homolog 4 lysine demethylase alkB homolog 4 lysine demthylase alkB alkylation repair homolog 4 alkB alkylation repair homolog 4 (E. coli) ALKBH4 Alkylated DNA repair protein alkB homolog 4 Alpha-ketoglutarate-dependent dioxygenase alkB homolog 4 DNA N6-methyl adenine demethylase ALKBH4 Lysine-specific demethylase ALKBH4 probable alpha-ketoglutarate-dependent dioxygenase ABH4 RGD1308608; alkB homolog 4, lysine demthylase; alkB, alkylation repair homolog 4; Alkylated DNA repair protein alkB homolog 4; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 4; DNA N6-methyl adenine demethylase ALKBH4; Lysine-specific demethylase ALKBH4; probable alpha-ketoglutarate-dependent dioxygenase ABH4
Gene Aliases: 2010004B12Rik; ABH4; Abj4; ALKBH4
UniProt ID: (Human) Q9NXW9, (Mouse) Q9D8F1
Entrez Gene ID: (Human) 54784, (Mouse) 72041
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.