Invitrogen
Manufacturer Code:PA576827
Catalog # PIPA576827
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human ALKBH3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABH3 AlkB homolog 3 alkB alkylation repair homolog 3 (E. coli) ALKB3&lowbarHUMAN ALKBH3 Alkylated DNA repair protein alkB homolog 3 alkylation repair homolog 3 Alpha-ketoglutarate-dependent dioxygenase alkB homolog 3 DEPC 1 DEPC-1 DEPC1 EC 1.14.11. FLJ43614 mABH3 MGC118790 MGC118792 MGC118793 MGC134125 PCA1 Prostate cancer antigen 1 prostate cancer antigen-1 RP23-375N21.4; AlkB homolog 3; alkB homolog 3, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 3; Alkylated DNA repair protein alkB homolog 3; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 3; DEPC-1; hABH3; mAbh3; Prostate cancer antigen 1; prostate cancer antigen-1
Gene Aliases: 1700108H04Rik; 1810020C19Rik; ABH3; ALKBH3; DEPC-1; DEPC1; hABH3; mABH3; PCA1
UniProt ID: (Human) Q96Q83, (Rat) Q5XIC8, (Mouse) Q8K1E6
Entrez Gene ID: (Human) 221120, (Rat) 362169, (Mouse) 69113
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.