Invitrogen
Manufacturer Code:PA565438
Catalog # PIPA565438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human ALKBH3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alkB homolog 3, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 3; Alkylated DNA repair protein alkB homolog 3; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 3; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 3 11411- Alkylated DNA repair protein alkB homolog 3 DEPC-1 Prostate cancer antigen 1 ALKBH3 ABH3 DEPC1; DEPC-1; hABH3; Prostate cancer antigen 1; prostate cancer antigen-1
Gene Aliases: ABH3; ALKBH3; DEPC-1; DEPC1; hABH3; PCA1
UniProt ID: (Human) Q96Q83
Entrez Gene ID: (Human) 221120
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.