Novus Biologicals
Manufacturer Code:NBP184274
Catalog # NBP184274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:WAAPVKSQAIAQPATTAKSHLHQKPGQTWKNKEHHLSDREFVFKEPQQVVRRAPEPRVIDREGVYEISLSPTGVSRVCLYPGFVDVKEADWILEQLCQDVPWKQRTGIREDITYQQPRLTAWYGEL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABH3MGC118793 alkB alkylation repair homolog 3 (E. coli) Alkylated DNA repair protein alkB homolog 3 DEPC1alpha-ketoglutarate-dependent dioxygenase alkB homolog 3 DEPC-1MGC118792 EC 1.14.11 EC 1.14.11.- FLJ43614 MGC118790 PCA1 Prostate cancer antigen 1 prostate cancer antigen-1; alkB homolog 3, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 3; Alkylated DNA repair protein alkB homolog 3; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 3; DEPC-1; hABH3; Prostate cancer antigen 1; prostate cancer antigen-1
Gene Aliases: ABH3; ALKBH3; DEPC-1; DEPC1; hABH3; PCA1
UniProt ID: (Human) Q96Q83
Entrez Gene ID: (Human) 221120
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.