Proteintech
Manufacturer Code:122921AP150UL
Catalog # 50-172-7865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALKBH3 Fusion Protein Ag2982 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABH3 ALKBH3 DEPC 1 DEPC1 PCA1 Prostate cancer antigen 1; alkB homolog 3, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 3; Alkylated DNA repair protein alkB homolog 3; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 3; DEPC-1; hABH3; mAbh3; Prostate cancer antigen 1; prostate cancer antigen-1
Gene Aliases: 1700108H04Rik; 1810020C19Rik; ABH3; ALKBH3; DEPC-1; DEPC1; hABH3; mABH3; PCA1
UniProt ID: (Human) Q96Q83, (Mouse) Q8K1E6
Entrez Gene ID: (Human) 221120, (Mouse) 69113
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.