Invitrogen
Manufacturer Code:PA5113048
Catalog # PIPA5113048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human DNA oxidative demethylase ALKBH2 protein (1-261AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2OG-Fe(II) oxy DC1; 2OG-Fe(II) oxy DC1 9530023G02 ABH2 AlkB homolog 2 alkB homolog 2 alpha-ketoglutarate dependent dioxygenase alkB homolog 2 alpha-ketoglutarate-dependent dioxygenase alkB alkylation repair homolog 2 alkB alkylation repair homolog 2 (E. coli) ALKBH2 Alkylated DNA repair protein alkB homolog 2 Alpha-ketoglutarate-dependent dioxygenase alkB homolog 2 AU016977 DNA oxidative demethylase ALKBH2 mABH2 Oxy DC1 RGD1306377; alkB homolog 2, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 2; Alkylated DNA repair protein alkB homolog 2; Alpha-ketoglutarate-dependent dioxygenase alkB homolog 2; DNA oxidative demethylase ALKBH2; Oxy DC1
Gene Aliases: ABH2; ALKBH2
UniProt ID: (Human) Q6NS38
Entrez Gene ID: (Human) 121642
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.