R&D Systems
Manufacturer Code:MAB7749
Catalog # MAB7749
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 810510 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human ALK-7 Leu26-Glu113 Accession # Q8NER5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type IC; activin A receptor type IC Activin receptor type IC activin receptor type-1C Activin receptor-like kinase 7 Activin RIC ACTR-IC ACVR1C ACVRLK7 ALK7 ALK7ALK-7 EC 2.7.11 EC 2.7.11.30; activin A receptor, type IC; Activin receptor type IC; Activin receptor type-1C; Activin receptor-like kinase 7; ACTR-IC; ACVR1C; ALK-7
Gene Aliases: ACVR1C; ACVRLK7; ALK7
UniProt ID: (Human) Q8NER5
Entrez Gene ID: (Human) 130399
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.