Novus Biologicals
Manufacturer Code:NBP130982
Catalog # NBP130982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ACVRL1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type II-like 1; activin A receptor type II-like 1 activin A receptor type II-like kinase 1 Activin receptor-like kinase 1 ACVRLK1ORW2 ALK-1 ALK1TGF-B superfamily receptor type I EC 2.7.11 HHT HHT2EC 2.7.11.30 serine/threonine-protein kinase receptor R3 SKR3 TSR-I; activin A receptor type IL; activin A receptor, type II-like kinase 1; Activin receptor-like kinase 1; ALK-1; Serine/threonine-protein kinase receptor R3; SKR3; TGF-B superfamily receptor type I; TSR-I
Gene Aliases: ACVRL1; ACVRLK1; ALK-1; ALK1; HHT; HHT2; ORW2; SKR3; TSR-I
UniProt ID: (Human) P37023
Entrez Gene ID: (Human) 94
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.