Invitrogen
Manufacturer Code:PA569321
Catalog # PIPA569321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat ALG8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: asparagine-linked glycosylation 8 alpha-13-glucosyltransferase-like protein; asparagine-linked glycosylation 8 homolog; asparagine-linked glycosylation 8 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8 homolog (yeast, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8, alpha-1,3-glucosyltransferase homolog; Asparagine-linked glycosylation protein 8 homolog; Dol-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichyl alpha-1,3-glucosyltransferase; dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase; dolichyl-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichol alpha- 1->3-glucosyltransferase; Dolichyl-P-Glc:Glc1Man9GlcNAc2-PP-dolichyl glucosyltransferase; dolichyl-P-glucose:Glc1Man9GlcNAc2-PP-dolichyl-alpha-1,3-glucosyltransferase; HUSSY-02; Probable dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1 3-glucosyltransferase Asparagine-linked glycosylation protein 8 homolog Dol-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichyl alpha-1 3-glucosyltransferase Dolichyl-P-Glc:Glc1Man9GlcNAc2-PP-dolichyl glucosyltransferase ALG8; Probable dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase; putative dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase-like protein
Gene Aliases: AI447372; ALG8; CDG1H; Gm1089; HUSSY-02; si:rp71-1p14.2; wu:fi19e09; wu:fi19f07; zgc:112507
UniProt ID: (Human) Q9BVK2, (Bovine) Q0P5D9, (Mouse) Q6P8H8
Entrez Gene ID: (Human) 79053, (Dog) 476793, (Pig) 100511030, (Bovine) 538731, (Rabbit) 100350058, (Horse) 100051152, (Mouse) 381903, (Guinea pig) 100732933, (Rat) 293129, (Zebrafish) 327601
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.