Novus Biologicals
Manufacturer Code:NBP169028
Catalog # NBP169028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to Alg8 (asparagine-linked glycosylation 8 alpha-13-glucosyltransferase homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of Alg8 (NP_001029299). Peptide sequence: LELKLLDPSQIPRASMTSGLVQQS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-13-glucosyltransferase) asparagine-linked glycosylation 8 homolog (S. cerevisiae asparagine-linked glycosylation 8 homolog (yeast asparagine-linked glycosylation 8 alpha-13-glucosyltransferase homolog (S.cerevisiae) Asparagine-linked glycosylation protein 8 homolog CDG1H dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-13-glucosyltransferase Dolichyl-P-Glc:Glc1Man9GlcNAc2-PP-dolichyl glucosyltransferase dolichyl-P-glucose:Glc1Man9GlcNAc2-PP-dolichyl-alpha-13-glucosyltransferase EC 2.4.1 EC 2.4.1.- HUSSY-02 MGC2840 probable dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-13-glucosyltransferase; asparagine-linked glycosylation 8 alpha-13-glucosyltransferase-like protein; asparagine-linked glycosylation 8 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8 homolog (yeast, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8, alpha-1,3-glucosyltransferase homolog; Asparagine-linked glycosylation protein 8 homolog; Dol-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichyl alpha-1,3-glucosyltransferase; dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase; dolichyl-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichol alpha- 1->3-glucosyltransferase; Dolichyl-P-Glc:Glc1Man9GlcNAc2-PP-dolichyl glucosyltransferase; dolichyl-P-glucose:Glc1Man9GlcNAc2-PP-dolichyl-alpha-1,3-glucosyltransferase; HUSSY-02; Probable dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase
Gene Aliases: ALG8; CDG1H; HUSSY-02
UniProt ID: (Human) Q9BVK2
Entrez Gene ID: (Human) 79053
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.