Invitrogen
Manufacturer Code:PA5113830
Catalog # PIPA5113830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal region of human ALG6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALG6 ALG6 alpha-13-glucosyltransferase ALG6 alpha-13-glucosyltransferase asparagine-linked glycosylation 6 (alpha-13-glucosyltransferase) asparagine-linked glycosylation 6 homolog (S. cerevisiae alpha-13-glucosyltransferase) asparagine-linked glycosylation 6 homolog (S. cerevisiae alpha-13-glucosyltransferase)-like asparagine-linked glycosylation 6 homolog (yeast alpha-13-glucosyltransferase) asparagine-linked glycosylation 6 homolog (yeast alpha-13-glucosyltransferase) asparagine-linked glycosylation 6 alpha-13-glucosyltransferase homolog asparagine-linked glycosylation 6 alpha-13-glucosyltransferase homolog (S. cerevisiae) Asparagine-linked glycosylation protein 6 Asparagine-linked glycosylation protein 6 homolog BOS_3497 CDG1C Dolichyl pyrophosphate Man9GlcNAc2 alpha-13-glucosyltransferase dolichyl-P-Glc:Man(9)GlcNAc(2)-PP-dolichol alpha- 1->3-glucosyltransferase dolichyl-P-Glc:Man(9)GlcNAc(2)-PP-dolichol alpha-13-glucosyltransferase Dolichyl-P-Glc:Man9GlcNAc2-PP-dolichyl glucosyltransferase dolichyl-P-Glc:Man9GlcNAc2-PP-dolichylglucosyltransferase Dol-P-Glc:Man(9)GlcNAc(2)-PP-Dol alpha-13-glucosyltransferase E230028F23Rik LOC100357591 Man(9)GlcNAc(2)-PP-Dol alpha-13-glucosyltransferase My046 UNA544 YOR002W zgc:100813; asparagine-linked glycosylation 6 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 6 homolog (yeast, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 6, alpha-1,3-glucosyltransferase homolog; Asparagine-linked glycosylation protein 6 homolog; Dol-P-Glc:Man(9)GlcNAc(2)-PP-Dol alpha-1,3-glucosyltransferase; Dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase; dolichyl-P-Glc:Man(9)GlcNAc(2)-PP-dolichol alpha- 1->3-glucosyltransferase; Dolichyl-P-Glc:Man9GlcNAc2-PP-dolichyl glucosyltransferase; dolichyl-P-Glc:Man9GlcNAc2-PP-dolichylglucosyltransferase; Man(9)GlcNAc(2)-PP-Dol alpha-1,3-glucosyltransferase
Gene Aliases: ALG6; CDG1C; My046
UniProt ID: (Human) Q9Y672
Entrez Gene ID: (Human) 29929
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.