Invitrogen
Manufacturer Code:PA5114185
Catalog # PIPA5114185
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: ALSYLEKRQK RDPAFTYEVI VVDDGSKDQT SKVAFKYCQK YGSDKVRVIT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1500026A19Rik 2600005J22Rik ALG5 Alg5 homolog ALG5 dolichyl-phosphate beta-glucosyltransferase Alg5 S. cerevisiae homolog of asparagine-linked glycosylation 5 (dolichyl-phosphate beta-glucosyltransferase) asparagine-linked glycosylation 5 homolog (S. cerevisiae dolichyl-phosphate beta-glucosyltransferase) asparagine-linked glycosylation 5 homolog (yeast dolichyl-phosphate beta-glucosyltransferase) asparagine-linked glycosylation 5 dolichyl-phosphate beta-glucosyltransferase homolog Asparagine-linked glycosylation protein 5 homolog AV009521 AV170001 AW545564 bA421P11.2 dolichyl phosphate glucosyltransferase Dolichyl-phosphate beta-glucosyltransferase DolP-glucosyltransferase HSPC149; Alg5, S. cerevisiae, homolog of; asparagine-linked glycosylation 5 homolog (S. cerevisiae, dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5 homolog (yeast, dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5, dolichyl-phosphate beta-glucosyltransferase homolog; Asparagine-linked glycosylation protein 5 homolog; dolichyl phosphate glucosyltransferase; Dolichyl-phosphate beta-glucosyltransferase; DolP-glucosyltransferase
Gene Aliases: ALG5; bA421P11.2; HSPC149
UniProt ID: (Human) Q9Y673
Entrez Gene ID: (Human) 29880
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.