Novus Biologicals
Manufacturer Code:NBP286978
Catalog # NB034600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of ALG5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alg5 S. cerevisiae homolog of asparagine-linked glycosylation 5 homolog (S. cerevisiae dolichyl-phosphatebeta-glucosyltransferase) asparagine-linked glycosylation 5 homolog (yeast dolichyl-phosphatebeta-glucosyltransferase) asparagine-linked glycosylation 5 dolichyl-phosphate beta-glucosyltransferasehomolog (S. cerevisiae) Asparagine-linked glycosylation protein 5 homolog bA421P11.2 dolichyl phosphate glucosyltransferase dolichyl-phosphate beta-glucosyltransferase DolP-glucosyltransferase EC 2.4.1 EC 2.4.1.117 RP11-421P11.2; Alg5, S. cerevisiae, homolog of; asparagine-linked glycosylation 5 homolog (S. cerevisiae, dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5 homolog (yeast, dolichyl-phosphate beta-glucosyltransferase); asparagine-linked glycosylation 5, dolichyl-phosphate beta-glucosyltransferase homolog; Asparagine-linked glycosylation protein 5 homolog; dolichyl phosphate glucosyltransferase; Dolichyl-phosphate beta-glucosyltransferase; DolP-glucosyltransferase
Gene Aliases: ALG5; bA421P11.2; HSPC149
UniProt ID: (Human) Q9Y673
Entrez Gene ID: (Human) 29880
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.