Invitrogen
Manufacturer Code:PA543263
Catalog # PIPA543263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ALG2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-1,3-mannosyltransferase ALG2; Alpha-1,3/1,6-mannosyltransferase ALG2; asparagine-linked glycosylation 2 (alpha-1,3-mannosyltransferase); Asparagine-linked glycosylation 2 alpha-1 3-mannosyltransferase homolog (S. cerevisiae) CDGIi FLJ14511 hALPG2 NET38; asparagine-linked glycosylation 2 homolog (S. cerevisiae, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (yeast, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2, alpha-1,3-mannosyltransferase homolog; Asparagine-linked glycosylation protein 2 homolog; GDP-Man:Man(1)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase; GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase; GDP-Man:Man(2)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase; homolog of yeast ALG2
Gene Aliases: 1110018A23Rik; 1300013N08Rik; ALG2; ALPG2; CDGIi; CMS14; CMSTA3; hALPG2; im:7145131; MNCb-5081; NET38; UNQ666/PRO1298
UniProt ID: (Human) Q9H553, (Mouse) Q9DBE8
Entrez Gene ID: (Human) 85365, (Rabbit) 100353637, (Dog) 474780, (Bovine) 538899, (Horse) 100054840, (Goat) 102172999, (Mouse) 56737, (Guinea pig) 100715517, (Rat) 313231, (Zebrafish) 403068
Molecular Function:
carbohydrate phosphatase
glycosyltransferase
hydrolase
nucleotidyltransferase
phosphatase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.