Invitrogen
Manufacturer Code:PA5109379
Catalog # PIPA5109379
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 197-416 of human ALG2 (NP_149078.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110018A23Rik 1300013N08Rik ALG2 ALPG2 Alpha-13/16-mannosyltransferase ALG2 Asparagine-linked glycosylation protein 2 Asparagine-linked glycosylation protein 2 homolog BOS_9019 CDGIi CMS14 CMSTA3 FLJ14511 GDP-Man:Man(1)GlcNAc(2)-PP-Dol alpha-13-mannosyltransferase GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase GDP-Man:Man(2)GlcNAc(2)-PP-Dol alpha-16-mannosyltransferase hALPG2 im:7145131 MNCb-5081 NET38 UNQ666/PRO1298 YGL065C; alpha-1,3-mannosyltransferase ALG2; Alpha-1,3/1,6-mannosyltransferase ALG2; asparagine-linked glycosylation 2 homolog (S. cerevisiae, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2 homolog (yeast, alpha-1,3-mannosyltransferase); asparagine-linked glycosylation 2, alpha-1,3-mannosyltransferase homolog; Asparagine-linked glycosylation protein 2 homolog; GDP-Man:Man(1)GlcNAc(2)-PP-Dol alpha-1,3-mannosyltransferase; GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase; GDP-Man:Man(2)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase; homolog of yeast ALG2
Gene Aliases: 1110018A23Rik; 1300013N08Rik; ALG2; ALPG2; CDGIi; CMS14; CMSTA3; hALPG2; MNCb-5081; NET38; UNQ666/PRO1298
UniProt ID: (Human) Q9H553, (Mouse) Q9DBE8
Entrez Gene ID: (Human) 85365, (Mouse) 56737
Molecular Function:
carbohydrate phosphatase
glycosyltransferase
hydrolase
nucleotidyltransferase
phosphatase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.